VWR® Spec-Wipe® 4, Cleanroom Wipes
Supplier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
115-0036EA 99.8 GBP
115-0036 115-0039
VWR® Spec-Wipe® 4, Cleanroom Wipes
Wipes Cleanroom Wipes Dry Cleanroom Wipes
Excellent for cleaning rough, abrasive, or irregular surfaces, yet soft enough for delicate surfaces. Appropriate for pharmaceutical and biotechnology industries.
  • For critical process cleaning, and environments where low particle and fibre generation is critical
  • Made from 100% double-knit polyester, laundered in ISO Class 4 cleanroom to reduce particulate and extractable contaminants
  • Durable, double-knit allows rigorous cleaning, is also soft for delicate surfaces
  • Re-sealable pouches maintain product integrity, preserve cleanliness and saturation levels
More Product Information