Iron(II) fluoride, anhydrous 98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

11486.09 11486.18
11486.09EA 61.7 GBP
11486.09 11486.18
Iron(II) fluoride, anhydrous 98%
Iron (II) fluoride

Purity: 98% CAS No.: 7789-28-8 Molecular Formula: FeF2 Formula Weight: 93.84 Physical Form: Powder Density: 4.09 Appearance: White Melting Point: 1000° subl. Merck Reference: 14,4045 UN No.: 3260 MDL No.: MFCD00016093

Formula: F₂Fe
Boiling Pt: 1100 °C (1013 hPa)
Melting Pt: >1000 °C (subl.)
Density: 4.09
Storage Temperature: Ambient
MDL Number: MFCD00016093
CAS Number: 7789-28-8
EINECS: 232-155-7
UN: 3260
ADR: 8,II
Merck Index: 14,4045

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR