Indium ≥99.998% (metals basis), wire, Puratronic®, Ø 2.0 mm (0.08 in)

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

11461.BU 11461.G1
11461.BUEA 103 GBP
11461.BU 11461.G1
Indium ≥99.998% (metals basis), wire, Puratronic®, Ø 2.0 mm (0.08 in)
Indium

Purity: 99.998+% CAS No.: 7440-74-6 Physical Form: ≈22.95g/m Odour: Odourless MDL No.: MFCD00134048

Formula: In
Boiling Pt: 2000 °C (1013 hPa)
Melting Pt: 156 °C
Density: 7.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134048
CAS Number: 7440-74-6
EINECS: 231-180-0
Merck Index: 13,04971

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR