Cresol red sodium salt, Sigma-Aldrich®

Supplier: SIGMA ALDRICH MICROSCOPY
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Cresol Red sodium, Sodium 4-(3-(4-hydroxy-3-methylphenyl)-1,1-dioxido-3H-benzo[c][1,2]oxathiol-3-yl)-2-methylphenolate

M-Clarity™ quality level = MQ100

114480-25G 114480-5G
114480-25GEA 87.4 GBP
114480-25G 114480-5G
Cresol red sodium salt, Sigma-Aldrich®
Cresol red sodium salt
Formula: C₂₁H₁₇NaO₅S
Storage Temperature: Ambient
MDL Number: MFCD00001618
CAS Number: 62625-29-0
EINECS: 263-654-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aed6540c89293aac7cae31e3977278f8c265eb2a&vid=780d5587ae42cbcbdfa45432bdbc2ad55511ffe4&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR