Silver bromide ≥99.5%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

11425.06 11425.14 11425.22
11425.06EA 32.7 GBP
11425.06 11425.14 11425.22
Silver bromide ≥99.5%
Silver bromide

Purity: 99.9% CAS No.: 7785-23-1 Molecular Formula: AgBr Formula Weight: 187.78 Physical Form: Powder Density: 6.47 Appearance: Pale yellow Odour: Odourless Melting Point: 432° Solubility: Practically insoluble in water. Insoluble in alcohol and acids. Partially soluble in ammonia Merck Reference: 14,8506 UN No.: 3077 MDL No.: MFCD00003398

Formula: AgBr
Boiling Pt: 1300 °C (1013 hPa)
Melting Pt: 432 °C
Density: 6.47
Storage Temperature: Ambient
MDL Number: MFCD00003398
CAS Number: 7785-23-1
EINECS: 232-076-8
ADR: 9,III
Merck Index: 14,8506

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR