Silver nitrate 99,9 Metals basis ACS

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Lunar caustic, Silver mononitrate, Nitric acid silver salt

11414.06 11414.14 11414.22 11414.36 11414.62 11414.A1
11414.06EA 35.2 GBP
11414.06 11414.14 11414.22 11414.36 11414.62 11414.A1
Silver nitrate 99,9 Metals basis ACS
Silver nitrate

Purity: 99.9+% CAS No.: 7761-88-8 Molecular Formula: AgNO3 Formula Weight: 169.87 Physical Form: Crystalline Density: 4.352 Appearance: White Odour: Odourless Melting Point: 212° Solubility: Freely soluble in water, alcohol, ammonia water. Slightly soluble in ether Merck Reference: 14,8518 UN No.: 1479 MDL No.: MFCD00003414


Silver plating, photography, manufacturing of other silver compounds, mirrors, as an analytical lab reagent, in Colouring porcelain

Formula: AgNO₃
MW: 169.87 g/mol
Boiling Pt: 444
Melting Pt: 212 °C
Density: 4.352
Storage Temperature: Ambient
MDL Number: MFCD00003414
CAS Number: 7761-88-8
EINECS: 231-853-9
UN: 1479
ADR: 5.1 I
REACH: 01-2119513705-43
Merck Index: 14,8518
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f56d5ff35f4c0937041d9629e42aae69bf1987b3&vid=7ca52a55b378bddc27fb4a51ace3ae27a0451c75&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR