Tychem® 6000 AL TF640 00 Ventilated Coveralls
Supplier: DuPont
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
TF0640TGY00/S TF0640TGY00/3X TF0640TGY00/L TF0640TGY00/M TF0640TGY00/XL TF0640TGY00/2X
114-9615EA 372 GBP
114-9615 114-9620 114-9617 114-9616 114-9618 114-9619
Tychem® 6000 AL TF640 00 Ventilated Coveralls
Disposable Garments
Air-fed coverall with cuff elastication and over boot assembly, consisting of integrated dissipative inner boots attached to dissipative outer boots.
  • Ventilated coverall with stitched and overtaped seams
  • Fully enclosed over hood with clear front and crown see-through visor
  • Front closure with double flap hook and loop, single zipper closing to center, horizontal opening
  • Three exhaust valves; two at hips, one in hood back
  • Air distribution made of Tychem 6000 fabric
  • Emergency egress
  • Cuff elastication and elastic thumb loop on inner sleeve opening
  • Over boot
More Product Information