ESD safety shoes, lace-up, L10, Sensation Low
Supplier: ELTEN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
72106/37 72106/38 72106/39 72106/40 72106/41 72106/42 72106/43 72106/44 72106/45 72106/46 72106/47
114-7625EA 89.9 GBP
114-7625 114-7626 114-7627 114-7628 114-7629 114-7630 114-7631 114-7632 114-7633 114-7634 114-7635
ESD safety shoes, lace-up, L10, Sensation Low
Shoes and Boots
Vintage and casual style, environmental friendly manufactured ESD safety shoe made from hydrophobised textile material in jeans look and hydrophobised nubuck leather, a padded tongue and a low profile dual-density TPU/PU sole L10.
  • Fabric lining Bioactive
  • Replaceable insole; orthopaedic insoles possible
  • Full-length inlay sole ESD PRO grey
  • Protective steel toecap (200 J)
Ideal for use in areas where exposure to moisture is expected.
Colour: Black/grey
Certifications: EN 61340-5-1 (ESD); DGUV 112-191; EN ISO 20345; S2, SRC
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification ISO 14001