ESD insole, ESD-PRO
Supplier: ELTEN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
204054/35 204054/36 204054/37 204054/38 204054/39 204054/40 204054/41 204054/42 204054/43 204054/44 204054/45 204054/46 204054/47 204054/48 204054/49 204054/50 204054/51 204054/52
114-7001EA 5.4 GBP
114-7001 114-7000 114-7002 114-7003 114-7004 114-7005 114-7006 114-7007 114-7012 114-7014 114-7015 114-7013 114-7016 114-7017 114-7008 114-7009 114-7010 114-7011
ESD insole, ESD-PRO
Shoe Insoles
Antibacterial and anatomically shaped ESD insole.
  • Open cell structure of PU foam guarantees high moisture absorption
  • Activated carbon absorber reduces odour
  • Skin friendly materials improve the foot climate
For use with L10, Biomex (Renzo Biomex), Trainers, Sportics, Classic-Line, Men at Work, Safety-Grip, Business (Ronny), White. Suitable for ESD safety shoes (electrical resistance of 105 to 107 Ohm).
Colour: Black