Occupational shoes, clogs, rubber, 9200 to 9310
Supplier: Abeba
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
9200/37 9200/38 9200/39 9200/40 9200/41 9200/42 9300/38 9300/39 9300/40 9300/41 9250/47 9200/36 9200/43 9200/44 9200/45 9200/46 9200/47 9250/36 9250/37 9250/38 9250/39 9250/40 9250/41 9250/42 9250/43 9250/44 9250/45 9250/46 9252/36 9252/37 9252/38 9252/39 9252/40 9252/41 9252/42 9252/43 9252/44 9252/45 9252/46 9252/47 9300/36 9300/37 9300/42 9300/43 9300/44 9300/45 9300/46 9300/47 9310/36 9310/37 9310/38 9310/39 9310/40 9310/41 9310/42 9310/43 9310/44 9310/45 9310/46 9310/47
114-5240EA 78.4 GBP
114-5240 114-5241 114-5242 114-5243 114-5244 114-5245 114-5277 114-5278 114-5279 114-5280 114-5262 114-5239 114-5246 114-5247 114-5248 114-5249 114-5250 114-5251 114-5252 114-5253 114-5254 114-5255 114-5256 114-5257 114-5258 114-5259 114-5260 114-5261 114-5263 114-5264 114-5265 114-5266 114-5267 114-5268 114-5269 114-5270 114-5271 114-5272 114-5273 114-5274 114-5275 114-5276 114-5281 114-5282 114-5283 114-5284 114-5285 114-5286 114-5287 114-5288 114-5289 114-5290 114-5291 114-5292 114-5293 114-5294 114-5295 114-5296 114-5297 114-5298
Occupational shoes, clogs, rubber, 9200 to 9310
Shoes and Boots
Anti-static clogs made from perforated leather upper, a foldable, adjustable heel strap and a rubber sole with cross/circle profile, resistant to acids, bases (cleaning agents) and fuels.
  • Leather lining
  • Sanitised® treatment reduces odour and bacterial growth
  • PU midsole
  • Slip resistant special rubber sole
Colour: White, black or blue.
Certifications: EN ISO 20347:2012; OB, FO, SRC
Order Now
SPECIFICATIONS
Environmentally Preferable