Insoles, Image
Supplier: HONEYWELL SAFETY
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
6000301/36 6000301/37 6000301/38 6000301/40 6000301/41 6000301/42 6000301/46 6000301/44 6000301/47 6000301/43 6000301/45 6000301/39 6000301/35
114-0486EA 28.1 GBP
114-0486 114-0487 114-0488 114-0490 114-0491 114-0492 114-0496 114-0494 114-0497 114-0493 114-0495 114-0489 114-0535
Insoles, Image
Shoe Insoles
Colour: light green.
  • Non woven fibres, honeycomb structure, hypoallergenic
  • Impregnated with anti-fungal, antibacterial agent, neutralises perspiration
  • Combats the main germs responsible for mycoses and other infection risks