Strontium titanate ≥99% (metals basis)

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

11399.30 11399.A1
11399.30EA 41.3 GBP
11399.30 11399.A1
Strontium titanate ≥99% (metals basis)
Strontium titanate

Purity: 99+% CAS No.: 12060-59-2 Molecular Formula: SrTiO3 Formula Weight: 183.51 Physical Form: Powder Density: 4.810 Melting Point: 2060° Solubility: Insoluble in water and most solvents MDL No.: MFCD00049554


Used in electronics and in electrical insulation

Formula: O₃SrTi
Melting Pt: 2060 °C
Density: 4.810
Storage Temperature: Ambient
MDL Number: MFCD00049554
CAS Number: 12060-59-2
EINECS: 235-044-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR