VWR® Advanced SF Cleanroom Sleeve Protectors (Bulk)
Supplier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
113-1193EA 173 GBP
113-1193 113-1194 113-9995 113-9996
VWR® Advanced SF Cleanroom Sleeve Protectors (Bulk)
Disposable Garments
VWR® Advanced SF sleeve protectors are manufactured from spunbonded polypropylene fabric (SPP), coated with breathable polyethylene film.
  • Elasticated cuffs, with sonically-welded seams
  • Durable fabric with good breathability and water transmission rate
  • Significant fluid and particle barrier
  • Particle shedding: Level II Helmke drum classification
  • Soft, cloth-like fabric
  • Thumb loop versions are also available
Tested and manufactured in an ISO certified facility under stringent process controls. Lot-controlled and validated through independent laboratory testing.