Quantus® Protect Xtra, Cleanroom Coats with Zip Closure
Supplier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
113-9953EA 0 GBP
113-9953 113-9955 113-9954 113-9952 113-9956 113-9957 113-9958
Quantus® Protect Xtra, Cleanroom Coats with Zip Closure
Workwear
These coats are made from Protect Xtra fabric which is a further development of the Protect fabric consisting of 100% polyester weave with conductive yarn (warp). This fabric has been especially designed for use in critical areas (e.g. operating rooms, sterile environments). Thanks to the PTFE finish, the coats provide very good splash protection against liquids and other contamination. The fabric has good tensile strength and provides very good resistance to microbial penetration.
  • Lapped seams ensure well-sealed and secure seams
  • Two way front zip closure,
More Product Information