VWR® Basic, Overalls
Supplier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
113-9002EA 112 GBP
113-9002 113-9003 113-9004 113-9005 113-9006 113-9007 113-9008 113-9009
VWR® Basic, Overalls
Disposable Garments
These disposable overalls, with collar, are made of non woven, spunbonded polypropylene (SPP), a lightweight fabric that provides protection in non hazardous, dirty work environments.
  • Collar
  • Zip closure (one direction)
  • Elasticated wrists
  • Free from natural rubber latex
Weight: 40 g/m²
Packaging: 1 piece per bag, 50 bags per case.