VWR® Advanced ESD Bouffant Caps
Supplier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
113-8901EA 118 GBP
113-8901
VWR® Advanced ESD Bouffant Caps
Disposable Garments
These disposable caps are made from a non woven, spunbonded polypropylene (SPP) fabric, which protects against dirt, grime and certain dry particulates in non hazardous environments.
  • Free from natural rubber latex
  • Anti-static properties
Weight: 16 g/m²
Packaging: 100 pieces per bag, 10 bags per case.