VWR®, Chemical Resistant Overalls
Supplier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
113-8379EA 231 GBP
113-8379 113-8383 113-8384 113-8380 113-8381 113-8382
VWR®, Chemical Resistant Overalls
Disposable Garments
These robust yet lightweight overalls are made from an optimised non woven PE fabric. Its softer feel and open structure still offers adequate Type 5 and 6 barrier protection and remains permeable to both air and moisture vapour.
  • Two-piece hood, zip closure with flap
  • Elasticated wrists, ankles, face and waist (stitched-in)
  • Anti-static treatment (EN 1149-1) on outside
  • Stitched external seams in yellow for visual identification and differentiation
  • Provides protection against radioactive contamination (EN 1073-2)
Colour: White
Weight: <180 g