VWR® Basic, Bouffant Caps, Latex-Free
Supplier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
113-1459EA 63.1 GBP
113-1459 113-1460 113-8762 113-8877 113-8887 113-8886 113-8888 113-8874 113-8875 113-8876 113-8879 113-8882 113-8880 113-8881 113-8883 113-8878 113-8884 113-8885
VWR® Basic, Bouffant Caps, Latex-Free
Disposable Garments
These disposable caps are made from a non woven, spunbonded polypropylene (SPP) fabric, which protects against dirt, grime, and certain dry particulates, in non hazardous environments.
  • Free from natural rubber latex
  • Available in a wide range of colours
Weight: 10 g/m²
Packaging: 100 pieces per bag, 10 bags per case.