Trousers, Tyvek® 500, model PT31L0

Supplier: DuPont
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

TYPT31SWHLG005 TYPT31SWHMD005 TYPT31SWHXL005 TYPT31SWH2X005
113-0481EA 275 GBP
113-0481 113-0482 113-0483 113-0484
Trousers, Tyvek® 500, model PT31L0
Workwear
Specially designed for use with Tyvek® apparel, these trousers can help offer enhanced protection for body parts that are more exposed to hazardous substances. Tyvek® 500 trousers are composed of flash spun high density polyethylene, providing an ideal balance of protection, durability and comfort. Tyvek® is permeable to both air and water vapour, yet repels water-based liquids and aerosols. It offers an excellent barrier against fine particles and fibres (down to 1 µm in size).

  • Fabric is ultra-low linting
  • Without pockets
  • Elasticated waist, no elastic at
More Product Information
Order Now

SPECIFICATIONS

Model 500 PT31L0
Environmentally Preferable
Garment application Laboratory
Risk area Anti-static
Gender Unisex
Sleeve or leg length Long
Fabric Tyvek®
Weight 41 g/m²

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR