Clamp gaskets, HelixMark®
Supplier: Freudenberg Medical Europe GmbH
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
70-400-05 70-400-07 70-400-10 70-400-15 70-400-20 70-400-25 70-400-30 70-400-40 70-400-60
113-0316EA 391 GBP
113-0316 113-0317 113-0318 113-0319 113-0320 113-0321 113-0322 113-0323 113-0324
Clamp gaskets, HelixMark®
Seals Gaskets
Silicone, platinum-cured.
  • Manufactured according to GMP in a production facility registered by the FDA for the manufacture of medical products
  • Can be sterilised by autoclaving, by ethylene oxide and via gamma radiation within the user's validation cycles and sterilisation levels
  • Meet USP Class VI requirements. Material certification and lot traceability are included in each box
These biomedical/pharmaceutical grade clamp gaskets promote crevice-free and protrusion-free processing. Contain no contaminants or leachables.
ISO 13485
Certifications: ISO 13485