PUREZERO* HG5 SGX* Nitrile Gloves, Blue, Non Sterile
Supplier: Halyard
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
CLN92512LG CLN92512MD CLN92512SM CLN92512XL CLN92512XS
112-6502EA 299 GBP
112-6502 112-6503 112-6504 112-6505 112-6506
PUREZERO* HG5 SGX* Nitrile Gloves, Blue, Non Sterile
Gloves
PUREZERO* HG5 gloves feature an accelerator-free formulation that reduces the risk of allergies and skin irritation associated with accelerator chemicals in other nitrile gloves. Comfortable to wear, the HG5 gloves also protect workers with effective barrier protection against chemical splash, micro-organisms, and viruses.
  • High-quality, non-sterile nitrile formulation
  • Manage product contamination with low particle levels (max >0,5 μm) <2000 particles/cm² for blue gloves
  • For use in cleanroom ISO 5 and higher, Grade B/C/D
  • Accelorator-free
  • Tacky grip
More Product Information