Life science nitrile gloves, PUREZERO® LIMON®
Supplier: Halyard
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
LFS311LG LFS311MD LFS311SM LFS311XS
112-6278EA 30.3 GBP
112-6278 112-6277 112-6276 112-6275
Life science nitrile gloves, PUREZERO® LIMON®
Gloves
PUREZERO® life science nitrile gloves deliver on all fronts. They are tested against 57 chemicals, with 3 levels of colour-coded protection offering increasing thickness designed for life science applications. The SAFESKIN® manufacturing facility uses 92% renewable energy, dispensers are made with up to 85% recycled materials, and cases are made with up to 99% recycled pulp.
  • Barrier protection (57 chemicals, 14 chemotherapy drugs)
  • Good fit and comfort
  • Textured fingertips provide tactile agility
  • Beaded cuff allows for ease of donning
  • Not made with natural
More Product Information