Nitrile gloves, Showa 707HVO
Supplier: Showa
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
707HVO-10/XL 707HVO-11/XXL 707HVO-7/S 707HVO-8/M 707HVO-9/L
112-6105EA 34.2 GBP
112-6105 112-6106 112-6108 112-6109 112-6110
Nitrile gloves, Showa 707HVO
Gloves
The 707HVO gloves are a more environmentally-conscious choice, breaking down by 82% in just 386 days when tested in a laboratory.
  • Eco-friendly alternative to standard single-use gloves
  • Highly tactile and fit like a second skin
  • Increased visibility thanks to fluorescent orange colour
  • Rolled cuff for easier donning
  • Provide protection against grease, chemicals, and abrasion
  • Ideal for use when working in laboratories or food preparation zones
Recommended for: Chemical spray and treatment; coating preparation; food packing and analysis; pharmaceuticals andMore Product Information
Order Now
SPECIFICATIONS
Model 707HVO
Environmentally Preferable Sustainable Materials,Safer Human and Environmental Health,Sustainable Packaging
Ambidextrous / hand-specific Hand-specific
Disposable / reusable Reusable
Latex / latex-free Latex-free
Material Nitrile
Finish Bisque
Cuff style Beaded
EN norm EN 388:2016 (1000X); EN ISO 374-1:2016 Type A (JKLOPT); EN ISO 374-5:2016
Food contact approved +
Environmental certification Biodegradation test results acc. to the norm ASTM D5526. Green circle