Industrial gloves, oil resistant, 660 / 690
Supplier: Showa
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
690-L 690-XL 690-M 690-XXL 660/36-10/XL 660/36-11/XXL 660-10/XL 660-11/XXL 660-9/L
112-5882EA 2130 GBP
112-5882 112-5884 112-5883 112-5885 112-5875 112-5876 112-5877 112-5878 112-5879
Industrial gloves, oil resistant, 660 / 690
Gloves
These fully PVC coated cotton knit gloves provide good mechanical and chemical resistance. They are impermeable and therefore suitable for working in damp or greasy environments.
  • Rough finish over entire surface: Good grip, even when objects are wet
  • Antibacterial and anti-odour treatment
  • Anatomical shape, seamless knit: No irritation for the skin
  • Flexible and soft, absorbs perspiration
  • Version with extended bonded arm (690) provides protection for the entire arm
Ideal for use in the automotive industry, in metallurgy and in the maritime sector.