Hand protectors, Hot Hand®
Supplier: FUNAKOSHI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
KG-0000-01 KG-0000-02 KG-0000-03 KG-0000-04 KG-0000-06
112-5070EA 83.3 GBP
112-5070 112-1983 112-5071 112-1984 112-5072
Hand protectors, Hot Hand®
Hot Hand Protectors
These gripping mittens made from flexible silicone elastomer can be used to handle hot or cold objects and to carry them short distances. The shape only protects the inside of the hand ensuring good sensitivity. The thumb must be inserted in one of the two openings, the other fingers in the remaining opening for a good grasping effect.
  • Temperature range: −60 to 230 °C
  • Clean with soapy water and store at room temperature away from damp and light
  • Assist with grip and ensure good contact
Colour: White, red
EN 407, EN 511; Cat. II
Certifications: More Product Information