Cleanroom gloves, polychloroprene, DermaShield® 73-711 / 73-721
Supplier: Ansell
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
73-721/6 73-721/6.5 73-721/7 73-721/7.5 73-721/8 73-721/8.5 73-721/9 73-711/7.5 73-711/9 73-711/8.5 73-711/6 73-711/6.5 73-711/7 73-711/8
112-3800EA 770 GBP
112-3800 112-3801 112-3802 112-3803 112-3804 112-3805 112-3806 112-3791 112-3794 112-3793 112-3788 112-3789 112-3790 112-3792
Cleanroom gloves, polychloroprene, DermaShield® 73-711 / 73-721
Gloves
Hand-specific, ergonomically designed gloves with a beaded cuff. Ideal for use in laboratories, pharmaceutical, biotech and medical device manufacturing.
  • Sterile or non sterile, powder-free, bisque fingertips
  • Optimal combination of product and worker protection
  • Accelerator free: No allergies
  • Softness and flexibility provides greater comfort compared to natural rubber latex
  • High resistance to puncture and a wide range of chemicals
  • Specific external surface treatment and constant monitoring of grip values
The sterile gloves are suitable for use in ClassMore Product Information