Glove liners, BO900
Supplier: Showa
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
BO900-UNI BO900-XL
112-3483EA 38.9 GBP
112-3483 112-3484
Glove liners, BO900
Gloves
These seamless knitted gloves made of nylon and polyester are ideal for use as undergloves.
  • Low linting, lightweight
  • Optimal dexterity
  • Exceptional capacity to absorb perspiration
  • Can be washed at 40 °C using a neutral detergent
Suitable for use in the following industries: Electronic components, Quality Control and integrated circuits.
Certifications: Cat. I
Order Now
SPECIFICATIONS
Model BO900
Environmentally Preferable
Ambidextrous / hand-specific Hand-specific
Disposable / reusable Reusable