Industrial gloves, Polytril™ Air ³/₄
Supplier: HONEYWELL SAFETY
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
2232272/10 2232272/7 2232272/8 2232272/9
112-1533EA 0 GBP
112-1533 112-1534 112-1535 112-1536
Industrial gloves, Polytril™ Air ³/₄
Gloves
Black polyamide knitted glove with black foam nitrile coating on palm and fingers, an open back and an elasticated knitted wrist.
  • Seamless knit construction conforms to the natural shape of the hand for greater strength, durability and ventilation
  • Foamy coating and ventilated back optimise the glove's breathability and improves the grip
  • Nitrile coating also provides outstanding abrasion resistance and impermeability to oils
The glove is ideal for handling of fine products in a greasy, damp or dirty atmosphere.
Certifications: EN 388:2016 (4121X); Cat. II