Dispensers for sterile gloves
Supplier: S-CURVE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
SD-300-C SD-300A SD-350-C
112-0536EA 602 GBP
112-0536 113-0339 SCURSD-350-C
Dispensers for sterile gloves
Glove Dispensers and Glove Box Holders
These safety dispensers are designed to protect prepacked sterile gloves from contamination.
  • Easy access first in/first out usage
  • Amber version to protect from ozone degradation
  • Available in free-standing or wall-mountable versions