Lens cleaning tissues, Grade 105, Whatman™
Supplier: Whatman products (Cytiva)
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
2105-862 2105-918 2105-841
111-5001EA 132 GBP
111-5001 111-5002 111-5000
Lens cleaning tissues, Grade 105, Whatman™
Wipes Disposable Wipes and Towels Lens Papers
These cleaning tissues are suitable for use with lenses and any other optical surfaces made from glass, quartz or plastic.
  • Soft surface
  • Strong and highly absorbent
  • Chemically pure tissue free from silicones and other additives