Safety faceshields, Shape™
Supplier: JSP
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
5SHA20C 5SHA2GEN20C
111-2594EA 27.9 GBP
111-2594 111-2595
Safety faceshields, Shape™
Face Shields
A compact, comfortable, and lightweight faceshield with superior optical quality.
  • Perfect optical quality
  • 2,2 mm polycarbonate visor
  • Wide field of vision
  • Adjustable wheel ratchet headband
  • Compatible with prescription glasses
The Shape™ Safety Faceshield features a moulded polycarbonate visor that provides superior optical quality with an exceptional field of vision.
Certifications: EN 166; EN 170.