Safety spectacles, 3700 series, SecureFit™
Supplier: 3M
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
SF3707XSGAFBLU SF3702SGAF-BLU SF3701ASP-BLU SF3701SGAF-BLU SF3701SGAF-GRN SF3701SGAF-ORG SF3701XSGAFBLU SF3707SGAF-BLU SF3702XSGAFBLU SF3703SGAF-GRN
111-2529EA 26.8 GBP
111-2529 111-2525 111-2520 111-2521 111-2522 111-2523 111-2524 111-2528 111-2526 111-2527
Safety spectacles, 3700 series, SecureFit™
Safety Glasses
SecureFit™ 3700 series are safety over-spectacles designed for use with prescription glasses. These lightweight over-spectacles are designed to fit over the users existing glasses to provide personalised protection.
  • Adjustable ratchet temples help to ensure a secure fit
  • Pressure Diffusion Temples (PDT) reduce pressure over the ear for a secure and comfortable fit
  • Polycarbonate lenses absorb 99,9% of UVA and UVB rays
  • Anti-scratch and anti-fog coatings provide increased durability and improved vision
  • Similar to typical sunglasses, grey lenses are popular in
More Product Information