Disposable earplugs, uvex xact-fit detec
Supplier: UVEX
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
2124.013 2124.011
111-1904EA 390 GBP
111-1904 111-1900
Disposable earplugs, uvex xact-fit detec
Earplugs Disposable Earplugs
The xact-fit detec is a detectable disposable earplug for use in production areas. The connecting cord makes it practical to wear and limits the risk of process contamination.
  • Design adapts perfectly to the shape of the ear reducing pressure on the ear canal, even when worn for long periods
  • Stems are marked for left and right ears giving improved wearer comfort and easy, accurate insertion and removal
  • Cord and stem reduce the potential for contamination as there is no need to touch the foam earplug before insertion
  • Blue colour makes the earplugs easier to
More Product Information