Optifit Twin Full Face Masks
Supplier: HONEYWELL SAFETY
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
1715231 1715241 1715251
DALL1715231EA 287 GBP
DALL1715231 111-1460 111-1461
Optifit Twin Full Face Masks
Respirators Reusable Respirators Full Face Respirators
Twin filter masks give general protection against particulates and/or toxic gases. They can only be used in sufficiently ventilated premises where the atmosphere contains a mininum oxygen volume of 17%.
  • Complete PC visor with wide field of vision
  • Facepiece seal on U-shaped silicone faceblank; lightweight and good facial compatibility (auto fit)
  • Five-strap head harness with quick-release system
  • Side lobes for wearing presciption spectacles inside mask
  • Built-in plastic speech diaphragm for better speech transmission (plastic membrane)
  • Exhalation valve made
More Product Information