Integrated suspension headcovers, Versaflo™ S-Series
Supplier: 3M
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
S657 S333LG S333SG S533L S533S S133L S133S S757 S655E
111-1118EA 296 GBP
111-1118 111-1110 111-1111 111-1112 111-1113 111-1108 111-1109 111-1119 111-2348
Integrated suspension headcovers, Versaflo™ S-Series
Respirators Respiratory Systems
The 3M™ S-series headcovers offer head and face protection, as well as providing eye and face protection from liquid splash and low energy impact (grade F). They are suitable for applications that require frequent replacement of the entire headgear assembly, such as pharmaceutical manufacturing and laboratory work, or in situations where there is less soiling of the outer fabric. All headcovers can be used in conjunction with 3M Powered and Supplied Air systems (e.g. Jupiter™ and Dustmaster™), thus offering respiratory protection.
  • Improved shape and visibility
  • Excellent airflow
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable