Versaflo™ M-300 Headtops for Respiratory System
Supplier: 3M
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
M306 M307
111-1086EA 592 GBP
111-1086 111-1087
Versaflo™ M-300 Headtops for Respiratory System
Respirators Respiratory Systems
Versaflo™ M-300 Series helmets offer respiratory, eye and face protection. In addition they incorporate head protection, essential for areas where hard hats are mandatory (e.g. construction sites).
  • Visor design combines excellent peripheral and downward vision with good optical clarity
  • Coated lenses provide chemical and scratch resistance
  • Helmet can be combined with a variety of spare parts and accessories
  • Optional hearing protection available
Certifications: EN 166, EN 397; M-307: EN12941 TH2, EN14594 2A.