Bayonet filters, X-plore® mask
Supplier: Dräger
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
6738038 6738873 6738872 6738817 6738816 6738775 6738776 6738778 6738039 6738874
111-1046EA 3.8 GBP
111-1046 111-1056 111-1055 111-0770 111-1054 111-1049 111-1052 111-2347 DRAE6738039 DRAE6738874
Bayonet filters, X-plore® mask
Respirators Reusable Respirators Respirator Filters
Range with a Dräger-specific bayonet connection that fits X-plore® 3300/3500 half masks and X-plore® 5500 full face masks.
  • High level of user safety, great fit under face shields and visors
  • Robust gas filter with a blue ABS plastic housing (boiling point >65 °C)
  • White particle filter housing for easy identification of P3 filters
  • Option to upgrade a gas filter to a combination filter by simply adding a P1 or P2 pad or Dräger X-plore® Pure filter
  • Safe, quick and easy bayonet connection with clear guidance for positioning the filter securely
  • Filters
More Product Information