Disposable Particle Filtering Respirators, FFP1/FFP2/FFP3, Classic
Supplier: Moldex
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
2405 236515 2400 2555 236015
111-0533EA 148 GBP
111-0533 111-0525 111-0532 111-1143 111-1142
Disposable Particle Filtering Respirators, FFP1/FFP2/FFP3, Classic
Respirators Disposable Respirators Particulate Respirators
Classic FFP masks protect against dust, mist and fumes.
  • Proven shape and durable FFP mask
  • DuraMesh® - strong and durable structure, keeps the mask in shape
  • ActivForm® - no nose clip needed
  • Loop strap for quick, simple and safe on and off
Certifications: EN 149:2001+A1:2009.