Safety spectacles, Contour
Supplier: Bollé Safety
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
CONTESP CONTPSI CONTPSF
111-0113EA 11.04 GBP
111-0113 111-0112 111-0514
Safety spectacles, Contour
Safety Glasses
Solidity, protection, design, comfort and lightness make Contour safety glasses a unique model.
  • Non-slip TIPGRIP temples
  • Non-slip bridge
  • Model with ESP (Extra Sensory Perception) lens: An innovative coating to reduce dazzle and the harmful effects of blue light
Weight: 21 g
Certifications: EN 166, (EN 170, EN 172)
Delivery information: Supplied with microfibre pouch.