Reusable half masks, 6000 and 7000
Supplier: 3M
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
6100S 6200M 6300L 7501 7502 7503 6212M 6223M 7523M
111-0061EA 61.3 GBP
111-0061 111-0062 111-0063 111-0065 111-0066 111-0067 111-0465 111-0466 111-2197
Reusable half masks, 6000 and 7000
Respirators Reusable Respirators Half Face Respirators
Reusable, lightweight and comfortable to wear. Incorporating a soft, hypoallergenic face piece material and central exhalation valve.
  • Uses 6000/2000 series bayonet filters
  • Low maintenance respirator can be disassembled, cleaned and reused many times
  • No replacement parts are necessary except filters and cartridges
EN 140
Certifications: EN 140
Ordering information: Supplied without filters, which are sold separately (except 111-0465, 111-0466).