Palladium(II) oxide monohydrate 99.9% (≥73% Pd) (metals basis)

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

11041.04 11041.09
11041.04EA 522 GBP
11041.04 11041.09
Palladium(II) oxide monohydrate 99.9% (≥73% Pd) (metals basis)
Palladium (II) oxide monohydrate

Purity: 99.9% CAS No.: 64109-12-2 Molecular Formula: PdO.H2O Formula Weight: 140.41 (122.40anhy) Physical Form: Powder Melting Point: dec. MDL No.: MFCD00011172

Formula: OPd·1H₂O
Melting Pt: Dec.
Storage Temperature: Ambient
MDL Number: MFCD00011172
CAS Number: 64109-12-2
EINECS: 215-218-3
Merck Index: 14,6993

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR