Palladium(II) chloride (≥59.0% Pd) ≥99.9% (metals basis)

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

11034.04 11034.09 11034.18 11034.22 11034.30 11034.02
11034.04EA 361 GBP
11034.04 11034.09 11034.18 11034.22 11034.30 11034.02
Palladium(II) chloride (≥59.0% Pd) ≥99.9% (metals basis)
Palladium (II) chloride

Purity: 99.9% CAS No.: 7647-10-1 Molecular Formula: PdCl2 Formula Weight: 177.31 Physical Form: Crystalline Density: 4.0 Appearance: Red-brown Odour: Odourless Melting Point: 675° dec. Solubility: Soluble in dilute mineral acids, aqueous metal halides Merck Reference: 14,6990 MDL No.: MFCD00003558


Catalyst precursor

Formula: PdCl₂
Melting Pt: 675 °C (dec.)
Density: 4.0
Storage Temperature: Ambient
MDL Number: MFCD00003558
CAS Number: 7647-10-1
EINECS: 231-596-2
UN: 3260
ADR: 8,III
Merck Index: 14,6990

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR