Gold ≥99.9985% (metals basis), wire, Premion®, Ø 2.0 mm (0.08 in)

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

10964.BQ 10964.BU
10964.BQEA 789 GBP
10964.BQ 10964.BU
Gold ≥99.9985% (metals basis), wire, Premion®, Ø 2.0 mm (0.08 in)
Gold

Purity: 99.9985% CAS No.: 7440-57-5 Physical Form: ≈60.6g/m Odour: Odourless MDL No.: MFCD00003436

Formula: Au
Boiling Pt: 2800 °C (1013 hPa)
Melting Pt: 1063 °C
Density: 19.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9
Merck Index: 12,04526

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR