Samarium(III) oxide ≥99.998% (metals basis), REacton® (REM ≤ 100 ppm)

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

10915.06 10915.14 10915.22
10915.06EA 47.8 GBP
10915.06 10915.14 10915.22
Samarium(III) oxide ≥99.998% (metals basis), REacton® (REM ≤ 100 ppm)
Samarium (III) oxide

Purity: 99.998% CAS No.: 12060-58-1 Molecular Formula: Sm2O3 Formula Weight: 348.70 Physical Form: Powder Refractive Index: 1.97 Melting Point: 2300° Merck Reference: 14,8347 MDL No.: MFCD00011237

Formula: O₃Sm₂
Melting Pt: 2300 °C
Density: 7.68 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011237
CAS Number: 12060-58-1
EINECS: 235-043-6
Merck Index: 14,8347

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR