Tin(II) chloride hydrate ≥99.995% (metals basis), Puratronic®

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Tin dichloride hydrate, Stannous chloride hydrate

10894.14 10894.06 10894.22
10894.14EA 160 GBP
10894.14 10894.06 10894.22
Tin(II) chloride hydrate ≥99.995% (metals basis), Puratronic®
Tin(II) chloride hydrate

Tin(II) chloride is widely used as a reducing agent in organic synthesis, for example conversion of quinones into hydroquinones, nitroaromatics into aromatic amines, and in Stephen reduction.


It can be used to reduce certain metal salts into metals, tin plating, analytical testing of mercury ions, formation of silver mirrors and metal-metal bonds. It is also used for tin-plating, as a mordant in textile dyeing and as a catalyst in the production of polylactic acid. Further it is used as a food additive, wherein it serves as an anti-oxidant and a color retention agent. It can be used in the condensation of aryl aldehydes with cyclohexan-1,3-diones to synthesize xanthenes. It catalyzes direct conversion of aldehydes into beta-keto esters.

Formula: SnCl₂·2H₂O
MW: 189.62 g/mol (anhy)
Melting Pt: 43…46 °C
Density: 2.71
Storage Temperature: Ambient
MDL Number: MFCD00149863
CAS Number: 10025-69-1
EINECS: 231-868-0
UN: 3260
ADR: 8 III
Merck Index: 14,8783

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR