Molybdenum(VI) oxide ≥99.998% (metals basis excluding W), Puratronic® (max. 300 ppm W)

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Molybdenum trioxide

10812.14 10812.22
10812.14EA 145 GBP
10812.14 10812.22
Molybdenum(VI) oxide ≥99.998% (metals basis excluding W), Puratronic® (max. 300 ppm W)
Molybdenum (VI) oxide

Purity: 99.998% CAS No.: 1313-27-5 Molecular Formula: MoO3 Formula Weight: 143.94 Physical Form: Powder Density: 4.692 Appearance: White to grey Odour: Odourless Melting Point: 795° Boiling Point: 1155° subl. Merck Reference: 14,6239 UN No.: 3288 MDL No.: MFCD00003469

Formula: MoO₃
Boiling Pt: 1155 °C (subl.)
Melting Pt: 795 °C
Density: 4.692
Storage Temperature: Ambient
MDL Number: MFCD00003469
CAS Number: 1313-27-5
EINECS: 215-204-7
UN: 3288
ADR: 6.1,III
Merck Index: 14,6239

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR