EDTA disodium salt dihydrate, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®

Supplier: Avantor
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Ethylenediaminetetraacetic acid, disodium salt, dihydrate, Ethylenediaminetetraacetic acid disodium salt dihydrate, Kestranal® 2S dihydrate, Disodium dihydrogen ethylenediaminetetraacetate dihydrate, Ethylenedinitrilotetra-acetic acid disodium salt dihydrate, Ethylenediaminetetraacetic acid disodium dihydrate, Titriplex III, Ethylenediaminetetra-acetic acid disodium salt dihydrate, Kestranal® 2S 2 H2O, Sequestrene Na₂, Edetate disodium dihydrate, Edetic acid disodium salt dihydrate

1073.0250EA 90 GBP
1073.0250 1073.0500 1073.1000
EDTA disodium salt dihydrate, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
EDTA disodium salt dihydrate
Formula: C₁₀H₁₄N₂Na₂O₈·2H₂O
Melting Pt: 255 °C
Storage Temperature: Ambient
MDL Number: MFCD00150037
CAS Number: 6381-92-6
EINECS: 205-358-3
Merck Index: 13,03543
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f89002087c5d4d84af5dded62c87287b1774fab&vid=0ef4c3febbcc8a20569606067799db31f2540748&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.0 - 101.0 %
Heavy Metals (as Pb) max. 0.001 %
Insoluble Matter max. 0.005 %
pH of 5% Solution at 25°C 4.0 - 6.0
Sulfate (SO4) max. 0.01 %
Trace Impurities (in ppm) Copper (Cu) max. 3
Trace Impurities (in ppm)Iron (Fe) max. 5


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR