Calcium oxide 99,998 Metals basis (excluding other alkaline earth and alkali metals 130 ppm max), Puratronic®

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Quicklime, Calcium oxide from marble, caustic, Lime

10684.04 10684.09 10684.18 10684.30
10684.04EA 47.3 GBP
10684.04 10684.09 10684.18 10684.30
Calcium oxide 99,998 Metals basis (excluding other alkaline earth and alkali metals 130 ppm max), Puratronic®
Calcium oxide

Purity: 99.998% CAS No.: 1305-78-8 Molecular Formula: CaO Formula Weight: 56.08 Physical Form: Powder Density: 3.37 Refractive Index: 1.8300 Sensitivity: Air and Moisture Sensitive Appearance: White Odour: Odourless Melting Point: 2572° Boiling Point: 2850° Merck Reference: 14,1686 MDL No.: MFCD00010911

Formula: CaO
MW: 56.08 g/mol
Boiling Pt: 2850 °C
Melting Pt: 2572 °C
Density: 3.37
Storage Temperature: Ambient
MDL Number: MFCD00010911
CAS Number: 1305-78-8
EINECS: 215-138-9
UN: 1910
ADR: 8 III
Merck Index: 14,1686

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR