Cadmium(II) nitrate tetrahydrate ≥99.999% (metals basis), Puratronic®

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

10663.06 10663.14 10663.22
10663.06EA 36.6 GBP
10663.06 10663.14 10663.22
Cadmium(II) nitrate tetrahydrate ≥99.999% (metals basis), Puratronic®
Cadmium (II) nitrate tetrahydrate

Purity: 99.999% CAS No.: 10022-68-1 Molecular Formula: Cd(NO3)2.4H2O Formula Weight: 308.47 (236.41anhy) Physical Form: Crystalline Density: 2.45 Melting Point: 59° Boiling Point: 132° Merck Reference: 14,1622 UN No.: 3087 MDL No.: MFCD00149626

Formula: CdN₂O₆·4H₂O
Boiling Pt: 132 °C
Melting Pt: 59 °C
Density: 2.45
Storage Temperature: Ambient
MDL Number: MFCD00149626
CAS Number: 10022-68-1
EINECS: 233-710-6
UN: 3087
ADR: 5.1,II
Merck Index: 14,1622

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR