Amersham™ Protran® Western Blotting Membranes, Nitrocellulose
Supplier: Cytiva
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
10600033 10600044 10600041 10600042 10600032 10600043 10600046 10600045 10600012 10600000 10600001 10600002 10600005 10600006 10600007 10600011 10600010 10600093 10600094 10600114 10600115 10600116 10600062 10600073 10600125 10600124
10600033.EA 306 GBP
10600033. 10600044. 10600041. 10600042. 10600032. 10600043. 10600046. 10600045. 10600012. 10600000. 10600001. 10600002. 10600005. 10600006. 10600007. 10600011. 10600010. 10600093. 10600094. 10600114. 10600115. 10600116. 10600062. 10600073. 10600125. GEHE10600124
Amersham™ Protran® Western Blotting Membranes, Nitrocellulose
Transfer Membranes
Amersham™ Protran® membranes are manufactured from 100% pure nitrocellulose to ensure the highest binding capacity possible. They are compatible with a variety of detection methods, including isotopic, chemiluminescent, colorimetric and fluorescent. No methanol pre-wetting step is required, making this the membrane of choice for proteins which prefer aqueous environments. Prior to transfer the membrane is simply wetted with water, and then placed in the transfer buffer. No other pretreatment steps are necessary.
  • Proprietary nitrocellulose formula gives a long shelf life of bound
More Product Information